Daidzein, 98%, Thermo Scientific Chemicals
Daidzein, 98%, Thermo Scientific Chemicals
Daidzein, 98%, Thermo Scientific Chemicals
Daidzein, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Daidzein, 98%, Thermo Scientific Chemicals
CAS: 486-66-8 | C15H10O4 | 254.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mg
100 mg
Packaging:
Glass bottle
Catalog number ACR328230250
Price (MYR)
731.00
Add to cart
Quantity:
25 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
731.00
Add to cart
Chemical Identifiers
CAS29094-61-9
IUPAC NameN-[2-(4-{[(cyclohexylcarbamoyl)amino]sulfonyl}phenyl)ethyl]-5-methylpyrazine-2-carboxamide
Molecular FormulaC21H27N5O4S
InChI KeyZJJXGWJIGJFDTL-UHFFFAOYSA-N
SMILESCC1=CN=C(C=N1)C(=O)NCCC1=CC=C(C=C1)S(=O)(=O)NC(=O)NC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Residue after ignition=<0.4 %
Appearance (Color)White to off-white
Loss on drying=<1 % (100°C, 3 hrs) (vacuum)
Heavy metals=<50 ppm