AEBSF hydrochloride, 98%
AEBSF hydrochloride, 98%
AEBSF hydrochloride, 98%
AEBSF hydrochloride, 98%
Thermo Scientific Chemicals
AEBSF hydrochloride, 98%
CAS: 30827-99-7 | C8H10FNO2S·ClH | 239.7 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
1 g
Packaging:
Glass bottle
Catalog number ACR328110500
Price (MYR)
562.00
Add to cart
Quantity:
50 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
562.00
Add to cart
Chemical Identifiers
CAS2576-47-8
IUPAC Namehydrogen 2-bromoethan-1-amine bromide
Molecular FormulaC2H7Br2N
InChI KeyWJAXXWSZNSFVNG-UHFFFAOYSA-N
SMILES[H+].[Br-].NCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals and/or chunks
Titration Argentometric>=98.5 %
Appearance (Color)White to light beige
Infrared spectrumConforms
Melting point172°C to 176°C