TMB QUICK, for ELISA, Liquid Substrate
TMB QUICK, for ELISA, Liquid Substrate
TMB QUICK, for ELISA, Liquid Substrate
TMB QUICK, for ELISA, Liquid Substrate
Thermo Scientific Chemicals
TMB QUICK, for ELISA, Liquid Substrate
Have Questions?
Change viewbuttonViewtableView
Substrate Quantity:
100mL
Catalog number ACR328051000
Price (MYR)
1,136.00
Add to cart
Substrate Quantity:
100mL
Request bulk or custom format
Price (MYR)
1,136.00
Add to cart
Chemical Identifiers
CAS7757-83-7
IUPAC Namedisodium heptahydrate sulfite
Molecular FormulaH14Na2O10S
InChI KeyWMJRPJZQQSSDBU-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.O.[Na+].[Na+].[O-]S([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
Assay (Redox Titration)≥97.5 to ≤102.5%
Identification (FTIR)Conforms
FormCrystalline powder or crystals