Xylenes, 99+%, for residue analysis, mixed, electronic use grade
Xylenes, 99+%, for residue analysis, mixed, electronic use grade
Xylenes, 99+%, for residue analysis, mixed, electronic use grade
Xylenes, 99+%, for residue analysis, mixed, electronic use grade
Thermo Scientific Chemicals
Xylenes, 99+%, for residue analysis, mixed, electronic use grade
CAS: 1330-20-7 | C8H10
Have Questions?
Catalog NumberQuantityPackaging
ACR3279500101 LGlass Bottle
Catalog number ACR327950010
Price (MYR)
585.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
585.00
Add to cart
Chemical Identifiers
CAS5194-32-1
IUPAC Name2-methylpyrimidine-5-carboxylic acid
Molecular FormulaC6H6N2O2
InChI KeyNMGIXZFBQPETOK-UHFFFAOYSA-N
SMILESCC1=NC=C(C=N1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Proton NMR>96.0%
For residue analysis
RUO – Research Use Only