CAPSO, 99%
CAPSO, 99%
CAPSO, 99%
Thermo Scientific Chemicals
CAPSO, 99%
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number ACR327741000
Price (MYR)
609.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
609.00
Add to cart
Specifications
Chemical Name or MaterialCAPSO
CAS73463-39-5
Absorbance(0.1M aq. soln.), 0.03 max. at 280nm, (0.1 M aq. soln.), (0.1 M aq. soln.) at 0nm
Melting Point263.0°C to 271.0°C
ColorWhite
View more
Biological buffer
RUO – Research Use Only