Diisopropyl azodicarboxylate, 94%, Thermo Scientific Chemicals
Diisopropyl azodicarboxylate, 94%, Thermo Scientific Chemicals
Diisopropyl azodicarboxylate, 94%, Thermo Scientific Chemicals
Diisopropyl azodicarboxylate, 94%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Diisopropyl azodicarboxylate, 94%, Thermo Scientific Chemicals
CAS: 2446-83-5 | C8H14N2O4 | 202.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number ACR327565000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialDiisopropyl azodicarboxylate
Name Note94%
CAS2446-83-5
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause damage to organs through prolonged or repeated exposure.
Toxic to aquatic life with long lasting effects.<br
View more
C-X Bond Formation (Halogen), Radical initiators
RUO – Research Use Only