Triton™ X-100, 98%, for molecular biology, DNAse, RNAse and Protease free
Triton™ X-100, 98%, for molecular biology, DNAse, RNAse and Protease free
Triton™ X-100, 98%, for molecular biology, DNAse, RNAse and Protease free
Triton™ X-100, 98%, for molecular biology, DNAse, RNAse and Protease free
Thermo Scientific Chemicals
Triton™ X-100, 98%, for molecular biology, DNAse, RNAse and Protease free
CAS: 9002-93-1 | C16H26O2, C16H26O2 | 250.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
250 mL
Packaging Type:
Glass bottle
Catalog number ACR327371000
Price (MYR)
584.00
Add to cart
Quantity:
100 mL
Packaging Type:
Glass bottle
Request bulk or custom format
Price (MYR)
584.00
Add to cart
Specifications
Assay98%
Chemical Name or MaterialTriton™ X-100
CAS9002-93-1
ColorUndesignated
Density1.0670 g/mL
View more
For molecular biology
RUO – Research Use Only