Toluene, 99.85%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Toluene, 99.85%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Toluene, 99.85%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Toluene, 99.85%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Toluene, 99.85%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Toluene, 99.85%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
Catalog number ACR326981000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS476620-54-9
IUPAC Name2-bromo-4,5-difluorobenzaldehyde
Molecular FormulaC7H3BrF2O
InChI KeyZHSZEWFWVDLMDL-UHFFFAOYSA-N
SMILESFC1=C(F)C=C(C=O)C(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%