1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
100 mL
1 L
Packaging:
View all
Glass Bottle
Catalog number ACR326930025
Price (MYR)
2,442.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
2,442.00
Add to cart
Chemical Identifiers
CAS4549-33-1
IUPAC Name1,9-dibromononane
Molecular FormulaC9H18Br2
InChI KeyWGAXVZXBFBHLMC-UHFFFAOYSA-N
SMILESBrCCCCCCCCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=96.0 %
Appearance (Color)Clear colorless to yellow
Refractive index1.4950 to 1.4980 (20°C, 589 nm)
Appearance (Form)Liquid
Suitable for battery materials development.
RUO – Research Use Only