n-Hexane, 96+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
n-Hexane, 96+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
n-Hexane, 96+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
n-Hexane, 96+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Hexane, 96+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Catalog number ACR326920025
Price (MYR)
1,514.00
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
1,514.00
Add to cart
Chemical Identifiers
CAS797755-07-8
IUPAC Name2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetic acid
Molecular FormulaC14H19BO4
InChI KeyFNLWHBHWDXCWHV-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1=CC=C(CC(O)=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Solid
Infrared spectrumConforms
Melting point162°C to 168°C
NMR (1H-NMR)Consistent with structure