n-Hexane, 96+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
n-Hexane, 96+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
n-Hexane, 96+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
n-Hexane, 96+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Hexane, 96+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Catalog number ACR326920010
Price (MYR)
796.00
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
796.00
Add to cart
Specifications
Chemical Name or Materialn-Hexane
Melting Point-95.0°C
CAS110-54-3
CAS Min %96.0