Ethyl acetate, 99.9%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Ethyl acetate, 99.9%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Ethyl acetate, 99.9%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Ethyl acetate, 99.9%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethyl acetate, 99.9%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Catalog number ACR326900010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS89379-79-3
IUPAC Name3-chloro-4-methoxypyridine
Molecular FormulaC6H6ClNO
InChI KeyOFJIIJJCVDJQFW-UHFFFAOYSA-N
SMILESCOC1=C(Cl)C=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%