Dichloromethane, 99.9%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals
Dichloromethane, 99.9%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals
Dichloromethane, 99.9%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals
Dichloromethane, 99.9%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dichloromethane, 99.9%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals
Dichloromethane, 99.9%, CH2Cl2, CAS Number-75-09-2, 513-35-9, aerothene mm, methylene chloride, freon 30, methylene bichloride, methane, dichloro, methane dichloride, narkotil, methylene dichloride, solmethine, solaesthin, 2.5L, 01, 60, 39.0 deg.C to 40.0 deg.C, CHEBI:15767, Colorless to Yellow
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
100 mL
1 L
Catalog number ACR326850025
Price (MYR)
1,091.00
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
1,091.00
Add to cart
Chemical Identifiers
CAS109-04-6
IUPAC Name2-bromopyridine
Molecular FormulaC5H4BrN
InChI KeyIMRWILPUOVGIMU-UHFFFAOYSA-N
SMILESBrC1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow or pale brown
FormLiquid
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Refractive Index1.5695-1.5725 @ 20?C