Dichloromethane, 99.9%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals
Dichloromethane, 99.9%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals
Dichloromethane, 99.9%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals
Dichloromethane, 99.9%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dichloromethane, 99.9%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals
Dichloromethane, 99.9%, CH2Cl2, CAS Number-75-09-2, 513-35-9, aerothene mm, methylene chloride, freon 30, methylene bichloride, methane, dichloro, methane dichloride, narkotil, methylene dichloride, solmethine, solaesthin, 2.5L, 01, 60, 39.0 deg.C to 40.0 deg.C, CHEBI:15767, Colorless to Yellow
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
100 mL
2.5 L
Catalog number ACR326850010
Price (MYR)
670.00
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
670.00
Add to cart
Specifications
Chemical Name or MaterialDichloromethane
Melting Point-97.0°C
CAS75-09-2
ColorColorless to Yellow
Health Hazard 1Warning