Chloroform, 99.9%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals
Chloroform, 99.9%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals
Chloroform, 99.9%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals
Chloroform, 99.9%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals
Chloroform, 99.9%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chloroform, 99.9%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals
CAS: 67-66-3 | CHCl3 | 119.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number ACR326821000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS1072-71-5
IUPAC Name1,3,4-thiadiazolidine-2,5-dithione
Molecular FormulaC2H2N2S3
InChI KeyBIGYLAKFCGVRAN-UHFFFAOYSA-N
SMILESS=C1NNC(=S)S1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to yellow
Appearance (Form)Powder
Infrared spectrumConforms
Melting point156°C to 162°C (Dec)
Titration with NaOH>=97.5 %