Acetonitrile, 99.9+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Acetonitrile, 99.9+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Acetonitrile, 99.9+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Acetonitrile, 99.9+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Acetonitrile, 99.9+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetonitrile, 99.9+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Acetonitrile, >-99.9%, C2H3N, CAS Number-75-05-8, methylkyanid, methane, cyano, methyl cyanide, mecn, methanecarbonitrile, ethanenitrile, ethyl nitrile, cyanure de methyl, cyanomethane, acetonitrile, 100mL, 0.002 % max., Vapor Pressure: 97mbar at 20 deg.C, 99.9+%, 02, 183, 81.0 deg.C to 82.0 deg.C
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Catalog number ACR326811000
Price (MYR)
695.00
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
695.00
Add to cart
Chemical Identifiers
CAS641-70-3
IUPAC Name4-nitro-1,3-dihydro-2-benzofuran-1,3-dione
Molecular FormulaC8H3NO5
InChI KeyROFZMKDROVBLNY-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=CC2=C1C(=O)OC2=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to pale brown or cream
FormCrystalline powder
Assay (Aqueous acid-base Titration)≥96.0%
Melting Point (clear melt)160-166°C