Petroleum ether, for residue analysis, 40° to 60°C, for trace anal. of polyarom. hydrocarbons, Thermo Scientific Chemicals
Petroleum ether, for residue analysis, 40° to 60°C, for trace anal. of polyarom. hydrocarbons, Thermo Scientific Chemicals
Petroleum ether, for residue analysis, 40° to 60°C, for trace anal. of polyarom. hydrocarbons, Thermo Scientific Chemicals
Petroleum ether, for residue analysis, 40° to 60°C, for trace anal. of polyarom. hydrocarbons, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Petroleum ether, for residue analysis, 40° to 60°C, for trace anal. of polyarom. hydrocarbons, Thermo Scientific Chemicals
CAS: 64742-49-0 | C6H14 | 86.178 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass Bottle
Catalog number ACR326790010
Price (MYR)
488.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
488.00
Add to cart
Chemical Identifiers
CAS76003-29-7
IUPAC Nametert-butyl 3-oxopiperazine-1-carboxylate
Molecular FormulaC9H16N2O3
InChI KeyFCMLWBBLOASUSO-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)N1CCNC(=O)C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)159.0-165.0?C
Assay (GC)≥97.5%
Appearance (Color)White to pale cream or pale yellow
For residue analysis
RUO – Research Use Only