n-Heptane, 99+%, residue analysis, for trace analysis of polyaromatic hydrocarb, Thermo Scientific Chemicals
n-Heptane, 99+%, residue analysis, for trace analysis of polyaromatic hydrocarb, Thermo Scientific Chemicals
n-Heptane, 99+%, residue analysis, for trace analysis of polyaromatic hydrocarb, Thermo Scientific Chemicals
n-Heptane, 99+%, residue analysis, for trace analysis of polyaromatic hydrocarb, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Heptane, 99+%, residue analysis, for trace analysis of polyaromatic hydrocarb, Thermo Scientific Chemicals
CAS: 142-82-5 | C7H16 | 100.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Catalog number ACR326770025
Price (MYR)
1,294.00
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
1,294.00
Add to cart
Chemical Identifiers
CAS128073-33-6
IUPAC Name2,2-difluoro-1-methylcyclopropane-1-carboxylic acid
Molecular FormulaC5H6F2O2
InChI KeyHLFLYOQLHYYNLT-UHFFFAOYNA-N
SMILESCC1(CC1(F)F)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Appearance (Color)White to yellow to pale brown
Melting Point (clear melt)60-66?C
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
For residue analysis
RUO – Research Use Only