n-Heptane, 99+%, residue analysis, for trace analysis of polyaromatic hydrocarb, Thermo Scientific Chemicals
n-Heptane, 99+%, residue analysis, for trace analysis of polyaromatic hydrocarb, Thermo Scientific Chemicals
n-Heptane, 99+%, residue analysis, for trace analysis of polyaromatic hydrocarb, Thermo Scientific Chemicals
n-Heptane, 99+%, residue analysis, for trace analysis of polyaromatic hydrocarb, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Heptane, 99+%, residue analysis, for trace analysis of polyaromatic hydrocarb, Thermo Scientific Chemicals
CAS: 142-82-5 | C7H16 | 100.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Catalog number ACR326770010
Price (MYR)
663.00
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
663.00
Add to cart
Chemical Identifiers
CAS133073-73-1
IUPAC Name(2S)-2-[(4-{[(2,4-diaminopteridin-6-yl)methyl](methyl)amino}phenyl)formamido]pentanedioic acid
Molecular FormulaC20H22N8O5
InChI KeyFBOZXECLQNJBKD-UGPWUYPHNA-N
SMILESCN(CC1=CN=C2N=C(N)N=C(N)C2=N1)C1=CC=C(C=C1)C(=O)N[C@@H](CCC(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Specific optical rotationon anhydrous substance
Appearance (Color)Yellow to orange
HPLC>=98.5 %
Specific optical rotation+19° to +24° (20°C, 589 nm) (c=1, 14 g/l Na2CO3)
View more
For residue analysis
RUO – Research Use Only