n-Heptane, 99+%, for residue analysis, ECD tested for pesticide analysis, Thermo Scientific Chemicals
n-Heptane, 99+%, for residue analysis, ECD tested for pesticide analysis, Thermo Scientific Chemicals
n-Heptane, 99+%, for residue analysis, ECD tested for pesticide analysis, Thermo Scientific Chemicals
n-Heptane, 99+%, for residue analysis, ECD tested for pesticide analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Heptane, 99+%, for residue analysis, ECD tested for pesticide analysis, Thermo Scientific Chemicals
CAS: 142-82-5 | C7H16 | 100.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Catalog number ACR326690025
Price (MYR)
1,118.00
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
1,118.00
Add to cart
Chemical Identifiers
CAS110-99-6
IUPAC Name2-(carboxymethoxy)acetic acid
Molecular FormulaC4H6O5
InChI KeyQEVGZEDELICMKH-UHFFFAOYSA-N
SMILESOC(=O)COCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with NaOH>=97.5 %
Appearance (Form)Crystalline powder
Melting point141°C to 145°C
Appearance (Color)White to light beige
For residue analysis
RUO – Research Use Only