n-Heptane, 99+%, for residue analysis, ECD tested for pesticide analysis, Thermo Scientific Chemicals
n-Heptane, 99+%, for residue analysis, ECD tested for pesticide analysis, Thermo Scientific Chemicals
n-Heptane, 99+%, for residue analysis, ECD tested for pesticide analysis, Thermo Scientific Chemicals
n-Heptane, 99+%, for residue analysis, ECD tested for pesticide analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Heptane, 99+%, for residue analysis, ECD tested for pesticide analysis, Thermo Scientific Chemicals
CAS: 142-82-5 | C7H16 | 100.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Catalog number ACR326690010
Price (MYR)
622.00
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
622.00
Add to cart
Chemical Identifiers
CAS99-56-9
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=97.5 %
Appearance (Color)Red-brown
Infrared spectrumConforms
HPLC>=97.5 %
Appearance (Form)Powder
For residue analysis
RUO – Research Use Only