Ethyl acetate, for residue analysis, suitable for residue and pesticides analysis, Thermo Scientific Chemicals
Ethyl acetate, for residue analysis, suitable for residue and pesticides analysis, Thermo Scientific Chemicals
Ethyl acetate, for residue analysis, suitable for residue and pesticides analysis, Thermo Scientific Chemicals
Ethyl acetate, for residue analysis, suitable for residue and pesticides analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethyl acetate, for residue analysis, suitable for residue and pesticides analysis, Thermo Scientific Chemicals
CAS: 141-78-6 | C4H8O2 | 88.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number ACR326610025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS7789-24-4
IUPAC Namelithium(1+) fluoride
Molecular FormulaFLi
InChI KeyPQXKHYXIUOZZFA-UHFFFAOYSA-M
SMILES[Li+].[F-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Total Metal Impurities≤0.01% (100 ppm)
FormPowder
For residue analysis
RUO – Research Use Only