Dichloromethane, for residue and pestic. anal., stab. with amylene, Thermo Scientific Chemicals
Have Questions?
Change view
Quantity:
2.5 L
1 L
Catalog number ACR326600025
Price (MYR)
598.00
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
598.00
Add to cart
Specifications
Chemical Name or MaterialDichloromethane
Melting Point-97.0°C
Name NoteECD Tested, For Pesticide analysis
CAS75-09-2
Health Hazard 1Warning