Dichloromethane, for residue and pestic. anal., stab. with amylene, Thermo Scientific Chemicals
Dichloromethane, for residue and pestic. anal., stab. with amylene, Thermo Scientific Chemicals
Dichloromethane, for residue and pestic. anal., stab. with amylene, Thermo Scientific Chemicals
Dichloromethane, for residue and pestic. anal., stab. with amylene, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dichloromethane, for residue and pestic. anal., stab. with amylene, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Catalog number ACR326600010
Price (MYR)
411.00
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
411.00
Add to cart
Chemical Identifiers
CAS15793-77-8
IUPAC Name2-hydrazinylquinoline
Molecular FormulaC9H9N3
InChI KeyQMVCLSHKMIGEFN-UHFFFAOYSA-N
SMILESNNC1=CC=C2C=CC=CC2=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale yellow to orange to red or brown
Assay from Supplier's CofA≥96.0% (HPLC) [U.S. Specification]
FormPowder or crystalline powder
Assay (GC)≥96.0% [U.K. Specification]
CommentMaterial Purchased in U.K. and U.S.
For residue analysis
RUO – Research Use Only