Hydroxylamine-O-sulfonic acid, 97%, Thermo Scientific Chemicals
Hydroxylamine-O-sulfonic acid, 97%, Thermo Scientific Chemicals
Hydroxylamine-O-sulfonic acid, 97%, Thermo Scientific Chemicals
Hydroxylamine-O-sulfonic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hydroxylamine-O-sulfonic acid, 97%, Thermo Scientific Chemicals
CAS: 2950-43-8 | H3NO4S | 113.09 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR32594025025 gGlass bottle
Catalog number ACR325940250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS68399-80-4
IUPAC Name3-[bis(2-hydroxyethyl)amino]-2-hydroxypropane-1-sulfonic acid
Molecular FormulaC7H17NO6S
InChI KeyXCBLFURAFHFFJF-UHFFFAOYNA-N
SMILESOCCN(CCO)CC(O)CS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)>98.5%