1,3,6,8-Pyrenetetrasulfonic acid tetrasodium salt, 85%, Thermo Scientific Chemicals
1,3,6,8-Pyrenetetrasulfonic acid tetrasodium salt, 85%, Thermo Scientific Chemicals
1,3,6,8-Pyrenetetrasulfonic acid tetrasodium salt, 85%, Thermo Scientific Chemicals
1,3,6,8-Pyrenetetrasulfonic acid tetrasodium salt, 85%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,3,6,8-Pyrenetetrasulfonic acid tetrasodium salt, 85%, Thermo Scientific Chemicals
CAS: 59572-10-0 | C16H6Na4O12S4 | 610.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number ACR325560010
Price (MYR)
677.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
677.00
Add to cart
Chemical Identifiers
CAS31106-82-8
IUPAC Name2-(bromomethyl)pyridine
Molecular FormulaC6H6BrN
InChI KeyOFPWMRMIFDHXFE-UHFFFAOYSA-N
SMILESBrCC1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pink to orange-brown
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Titration with HClO497.5 to 102.5 %
HPLC>=97.5 %