Urethane, 97%, Thermo Scientific Chemicals
Urethane, 97%, Thermo Scientific Chemicals
Urethane, 97%, Thermo Scientific Chemicals
Urethane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Urethane, 97%, Thermo Scientific Chemicals
CAS: 51-79-6 | C3H7NO2 | 89.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Packaging:
Plastic bottle
Catalog number ACR325540500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS2516-33-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4290 to 1.4330 (20°C, 589 nm)