Urethane, 97%, Thermo Scientific Chemicals
Urethane, 97%, Thermo Scientific Chemicals
Urethane, 97%, Thermo Scientific Chemicals
Urethane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Urethane, 97%, Thermo Scientific Chemicals
CAS: 51-79-6 | C3H7NO2 | 89.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
50 g
250 g
Packaging:
Plastic bottle
Catalog number ACR325540010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialUrethane
Name Note97%
CAS51-79-6
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful if swallowed.
May cause cancer.