Methyl sulfoxide-d6, for NMR, packaged in 0.50 ml ampoules, 99.9 atom % D
Methyl sulfoxide-d6, for NMR, packaged in 0.50 ml ampoules, 99.9 atom % D
Methyl sulfoxide-d6, for NMR, packaged in 0.50 ml ampoules, 99.9 atom % D
Methyl sulfoxide-d6, for NMR, packaged in 0.50 ml ampoules, 99.9 atom % D
Thermo Scientific Chemicals
Methyl sulfoxide-d6, for NMR, packaged in 0.50 ml ampoules, 99.9 atom % D
CAS: 2206-27-1 | C2H6OS | 84.17 g/mol
Have Questions?
Catalog NumberQuantity
ACR3253700505 mL
Catalog number ACR325370050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 mL
Request bulk or custom format
Chemical Identifiers
CAS286932-57-8
IUPAC Name2-bromo-1-fluoro-4-(trifluoromethoxy)benzene
Molecular FormulaC7H3BrF4O
InChI KeyVVVFPEJORKPEST-UHFFFAOYSA-N
SMILESFC1=C(Br)C=C(OC(F)(F)F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Refractive Index1.4475-1.4515 @ 20?C
For NMR
RUO – Research Use Only