Methanol-d4, for NMR, contains 0.03 v/v% TMS, 99.8% atom% D, AcroSeal™
Methanol-d4, for NMR, contains 0.03 v/v% TMS, 99.8% atom% D, AcroSeal™
Methanol-d4, for NMR, contains 0.03 v/v% TMS, 99.8% atom% D, AcroSeal™
Methanol-d4, for NMR, contains 0.03 v/v% TMS, 99.8% atom% D, AcroSeal™
Methanol-d4, for NMR, contains 0.03 v/v% TMS, 99.8% atom% D, AcroSeal™
Thermo Scientific Chemicals
Methanol-d4, for NMR, contains 0.03 v/v% TMS, 99.8% atom% D, AcroSeal™
CAS: 811-98-3 | CH4O | 36.066 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR325351000100 mLAcroSeal™ Glass Bottle
Catalog number ACR325351000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialMethanol-d4
Density0.8880g/mL
Deuterium (D)99.75% min.
Flash Point12°C
PackagingAcroSeal™ Glass Bottle
View more
For NMR
RUO – Research Use Only