Carbon tetrabromide, 98%, Thermo Scientific Chemicals
Carbon tetrabromide, 98%, Thermo Scientific Chemicals
Carbon tetrabromide, 98%, Thermo Scientific Chemicals
Carbon tetrabromide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Carbon tetrabromide, 98%, Thermo Scientific Chemicals
CAS: 558-13-4 | CBr4 | 331.64 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass Bottle
Catalog number ACR325150050
Price (MYR)
409.00
Add to cart
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
409.00
Add to cart
Chemical Identifiers
CAS22532-62-3
IUPAC Name4-bromo-2-hydroxybenzaldehyde
Molecular FormulaC7H5BrO2
InChI KeyHXTWKHXDFATMSP-UHFFFAOYSA-N
SMILESOC1=CC(Br)=CC=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>96.0%
C-X bond formation reagent
RUO – Research Use Only