(-)-Caryophyllene oxide, 95%, Thermo Scientific Chemicals
(-)-Caryophyllene oxide, 95%, Thermo Scientific Chemicals
(-)-Caryophyllene oxide, 95%, Thermo Scientific Chemicals
(-)-Caryophyllene oxide, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
(-)-Caryophyllene oxide, 95%, Thermo Scientific Chemicals
CAS: 1139-30-6 | C15H24O | 220.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number ACR325080500
Price (MYR)
796.00
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
796.00
Add to cart
Chemical Identifiers
CAS10152-76-8
IUPAC Name3-(methylsulfanyl)prop-1-ene
Molecular FormulaC4H8S
InChI KeyNVLPQIPTCCLBEU-UHFFFAOYSA-N
SMILESCSCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Refractive Index1.4700-1.4740 @ 20?C