Ethylbenzene-d10, for NMR, packaged in ampoules, 99+ atom% D
Ethylbenzene-d10, for NMR, packaged in ampoules, 99+ atom% D
Ethylbenzene-d10, for NMR, packaged in ampoules, 99+ atom% D
Ethylbenzene-d10, for NMR, packaged in ampoules, 99+ atom% D
Thermo Scientific Chemicals
Ethylbenzene-d10, for NMR, packaged in ampoules, 99+ atom% D
CAS: 25837-05-2 | C8H10 | 116.229 g/mol
Have Questions?
Catalog NumberQuantity
ACR3213600101 mL
Catalog number ACR321360010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialEthylbenzene-d10
Density0.9400g/mL
Deuterium (D)99.0% min.
Flash Point15°C
Physical FormLiquid
View more
For NMR
RUO – Research Use Only