Dichloromethane-d2, for NMR, packaged in 0.75 ml ampoules, 99.8 atom % D
Dichloromethane-d2, for NMR, packaged in 0.75 ml ampoules, 99.8 atom % D
Dichloromethane-d2, for NMR, packaged in 0.75 ml ampoules, 99.8 atom % D
Dichloromethane-d2, for NMR, packaged in 0.75 ml ampoules, 99.8 atom % D
Thermo Scientific Chemicals
Dichloromethane-d2, for NMR, packaged in 0.75 ml ampoules, 99.8 atom % D
CAS: 1665-00-5 | CH2Cl2 | 86.939 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR3207200757.5 mLAmpule
Catalog number ACR320720075
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
7.5 mL
Packaging:
Ampule
Request bulk or custom format
Chemical Identifiers
CAS176726-07-1
IUPAC Namescandium(3+) tris(trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane)
Molecular FormulaC6F18N3O12S6Sc
InChI KeyFUXLYEZEIZAKTL-UHFFFAOYSA-N
SMILES[Sc+3].FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
FormSolid
Assay from Suppliers CofA≥95.0%
For NMR
RUO – Research Use Only