Chloroform-d, for NMR, 100 atom % D, packaged in 0.75 ml ampoules
Chloroform-d, for NMR, 100 atom % D, packaged in 0.75 ml ampoules
Chloroform-d, for NMR, 100 atom % D, packaged in 0.75 ml ampoules
Chloroform-d, for NMR, 100 atom % D, packaged in 0.75 ml ampoules
Thermo Scientific Chemicals
Chloroform-d, for NMR, 100 atom % D, packaged in 0.75 ml ampoules
CAS: 865-49-6 | CHCl3 | 120.375 g/mol
Have Questions?
Catalog NumberQuantity
ACR3206800757.5 mL
Catalog number ACR320680075
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
7.5 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialChloroform-d
Density1.5000g/mL
Deuterium (D)99.95% min.
Physical FormLiquid
Water0.01% max. (Coulometric)
View more
For NMR
RUO – Research Use Only