Acetonitrile-d3, for NMR, packaged in 0.75 ml ampoules, 99.8 atom % D
Acetonitrile-d3, for NMR, packaged in 0.75 ml ampoules, 99.8 atom % D
Acetonitrile-d3, for NMR, packaged in 0.75 ml ampoules, 99.8 atom % D
Acetonitrile-d3, for NMR, packaged in 0.75 ml ampoules, 99.8 atom % D
Thermo Scientific Chemicals
Acetonitrile-d3, for NMR, packaged in 0.75 ml ampoules, 99.8 atom % D
CAS: 2206-26-0 | C2H3N | 44.07 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR3206600757.5 mLAmpule
Catalog number ACR320660075
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
7.5 mL
Packaging:
Ampule
Request bulk or custom format
Specifications
Chemical Name or MaterialAcetonitrile-d3
Density0.8440g/mL
Deuterium (D)99.5% min.
Flash Point2°C
PackagingAmpule
View more
For NMR
RUO – Research Use Only