Tris(dibenzylideneacetone)dipalladium(0), 97%, Thermo Scientific Chemicals
Tris(dibenzylideneacetone)dipalladium(0), 97%, Thermo Scientific Chemicals
Tris(dibenzylideneacetone)dipalladium(0), 97%, Thermo Scientific Chemicals
Tris(dibenzylideneacetone)dipalladium(0), 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tris(dibenzylideneacetone)dipalladium(0), 97%, Thermo Scientific Chemicals
CAS: 51364-51-3 | C51H42O3Pd2 | 915.73 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
5 g
Packaging:
Glass bottle
Catalog number ACR318775000
Price (MYR)
706.00
Add to cart
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
706.00
Add to cart
Chemical Identifiers
CAS104-75-6
IUPAC Name3-(aminomethyl)heptane
Molecular FormulaC8H19N
InChI KeyLTHNHFOGQMKPOV-UHFFFAOYNA-N
SMILESCCCCC(CC)CN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Refractive index1.4290 to 1.4320 (20°C, 589 nm)
Water=<0.2 %
GC>=98.5 %
Color scale=<20 APHA