Tris(dibenzylideneacetone)dipalladium(0), 97%, Thermo Scientific Chemicals
Tris(dibenzylideneacetone)dipalladium(0), 97%, Thermo Scientific Chemicals
Tris(dibenzylideneacetone)dipalladium(0), 97%, Thermo Scientific Chemicals
Tris(dibenzylideneacetone)dipalladium(0), 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tris(dibenzylideneacetone)dipalladium(0), 97%, Thermo Scientific Chemicals
CAS: 51364-51-3 | C51H42O3Pd2 | 915.73 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
500 mg
Packaging:
Glass bottle
Catalog number ACR318770050
Price (MYR)
2,298.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,298.00
Add to cart
Specifications
Chemical Name or MaterialTris(dibenzylideneacetone)dipalladium(0)
CAS51364-51-3
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
May cause an allergic skin reaction.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.