Palladium(II) trifluoroacetate, 97%, Thermo Scientific Chemicals
Palladium(II) trifluoroacetate, 97%, Thermo Scientific Chemicals
Palladium(II) trifluoroacetate, 97%, Thermo Scientific Chemicals
Palladium(II) trifluoroacetate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Palladium(II) trifluoroacetate, 97%, Thermo Scientific Chemicals
CAS: 42196-31-6 | C4F6O4Pd | 332.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number ACR317020010
Price (MYR)
1,200.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,200.00
Add to cart
Chemical Identifiers
CAS7051-16-3
IUPAC Name1-chloro-3,5-dimethoxybenzene
Molecular FormulaC8H9ClO2
InChI KeyWQHNWJBSROXROL-UHFFFAOYSA-N
SMILESCOC1=CC(OC)=CC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow or pale cream to brown
Melting Point (clear melt)31.0-38.0°C
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder or fused solid or clear liquid as melt