Rhenium pentacarbonyl chloride, 98%, Thermo Scientific Chemicals
Rhenium pentacarbonyl chloride, 98%, Thermo Scientific Chemicals
Rhenium pentacarbonyl chloride, 98%, Thermo Scientific Chemicals
Rhenium pentacarbonyl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Rhenium pentacarbonyl chloride, 98%, Thermo Scientific Chemicals
CAS: 14099-01-5 | C5ClO5Re | 361.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number ACR316310050
Price (MYR)
6,599.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
6,599.00
Add to cart
Chemical Identifiers
CAS58086-67-2
IUPAC Name2-fluoro-1-methylpyridin-1-ium 4-methylbenzene-1-sulfonate
Molecular FormulaC13H14FNO3S
InChI KeyHQWDKLAIDBOLFE-UHFFFAOYSA-M
SMILESC[N+]1=CC=CC=C1F.CC1=CC=C(C=C1)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay from Supplier's CofA≥88.0% (US-sourced material)
Elemental AnalysisC: 48.50-60.63% (Theory: 55.11%, UK-sourced material)
Elemental AnalysisN: 4.35-5.44% (Theory: 4.94%, UK-sourced material)
Assay (special)≥88.0% (based on elemental C, UK-sourced material)