Phosphorus pentoxide, 98+%, ACS reagent, Thermo Scientific Chemicals
Phosphorus pentoxide, 98+%, ACS reagent, Thermo Scientific Chemicals
Phosphorus pentoxide, 98+%, ACS reagent, Thermo Scientific Chemicals
Phosphorus pentoxide, 98+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phosphorus pentoxide, 98+%, ACS reagent, Thermo Scientific Chemicals
CAS: 1314-56-3 | O5P2 | 141.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number ACR315822500
Price (MYR)
494.00
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
494.00
Add to cart
Chemical Identifiers
CAS12125-02-9
IUPAC Nameammonium chloride
Molecular FormulaClH4N
InChI KeyNLXLAEXVIDQMFP-UHFFFAOYSA-N
SMILES[NH4+].[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric>=99.5 %
Heavy metals (as Pb)=<5 ppm
Sulfated ash=<0.01 %
Insoluble matter=<0.005 %
pH4.5 to 5.5 (5 % at 20°C)
View more
Adsorbent
RUO – Research Use Only