Boric acid, 99.99%, (trace metal basis), extra pure, Thermo Scientific Chemicals
Boric acid, 99.99%, (trace metal basis), extra pure, Thermo Scientific Chemicals
Boric acid, 99.99%, (trace metal basis), extra pure, Thermo Scientific Chemicals
Boric acid, 99.99%, (trace metal basis), extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Boric acid, 99.99%, (trace metal basis), extra pure, Thermo Scientific Chemicals
CAS: 10043-35-3 | BH3O3 | 61.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number ACR315185000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialBoric acid
ColorWhite
Name NoteExtra Pure
CAS10043-35-3
Melting Point169.0°C
View more
For ultra trace analysis, Lewis acid
RUO – Research Use Only