Boric acid, 99.99%, (trace metal basis), extra pure, Thermo Scientific Chemicals
Boric acid, 99.99%, (trace metal basis), extra pure, Thermo Scientific Chemicals
Boric acid, 99.99%, (trace metal basis), extra pure, Thermo Scientific Chemicals
Boric acid, 99.99%, (trace metal basis), extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Boric acid, 99.99%, (trace metal basis), extra pure, Thermo Scientific Chemicals
CAS: 10043-35-3 | BH3O3 | 61.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number ACR315181000
Price (MYR)
514.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
514.00
Add to cart
Chemical Identifiers
CAS539-74-2
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.4510 to 1.4540 (20°C, 589 nm)
GC>=98.5 %
Appearance (Color)Clear colorless to light yellow
For ultra trace analysis, Lewis acid
RUO – Research Use Only