Boric acid, 99.99%, (trace metal basis), extra pure, Thermo Scientific Chemicals
Boric acid, 99.99%, (trace metal basis), extra pure, Thermo Scientific Chemicals
Boric acid, 99.99%, (trace metal basis), extra pure, Thermo Scientific Chemicals
Boric acid, 99.99%, (trace metal basis), extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Boric acid, 99.99%, (trace metal basis), extra pure, Thermo Scientific Chemicals
CAS: 10043-35-3 | BH3O3 | 61.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Packaging:
Plastic bottle
Catalog number ACR315180025
Price (MYR)
3,764.00
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
3,764.00
Add to cart
Chemical Identifiers
CAS108-32-7
IUPAC Name4-methyl-1,3-dioxolan-2-one
Molecular FormulaC4H6O3
InChI KeyRUOJZAUFBMNUDX-UHFFFAOYNA-N
SMILESCC1COC(=O)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=99.4 %
Appearance (Color)Clear colorless
Infrared spectrumConforms
Water=<0.005 %
For ultra trace analysis, Lewis acid
RUO – Research Use Only