Chloroacetyl isocyanate, 90%, Tech., Thermo Scientific Chemicals
Chloroacetyl isocyanate, 90%, Tech., Thermo Scientific Chemicals
Chloroacetyl isocyanate, 90%, Tech., Thermo Scientific Chemicals
Chloroacetyl isocyanate, 90%, Tech., Thermo Scientific Chemicals
Acros Organics
Chloroacetyl isocyanate, 90%, Tech., Thermo Scientific Chemicals
CAS: 4461-30-7 | C3H2ClNO2 | 119.51 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number ACR314300050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS53721-12-3
IUPAC Name1,1'-diethyl-[4,4'-bipyridine]-1,1'-diium dibromide
Molecular FormulaC14H18Br2N2
InChI KeyLCEBDKLPALDQPV-UHFFFAOYSA-L
SMILES[Br-].[Br-].CC[N+]1=CC=C(C=C1)C1=CC=[N+](CC)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration Argentometric>=96.0 %
Appearance (Color)Yellow