Chloroacetyl isocyanate, 90%, Tech., Thermo Scientific Chemicals
Chloroacetyl isocyanate, 90%, Tech., Thermo Scientific Chemicals
Chloroacetyl isocyanate, 90%, Tech., Thermo Scientific Chemicals
Chloroacetyl isocyanate, 90%, Tech., Thermo Scientific Chemicals
Acros Organics
Chloroacetyl isocyanate, 90%, Tech., Thermo Scientific Chemicals
CAS: 4461-30-7 | C3H2ClNO2 | 119.51 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number ACR314300010
Price (MYR)
83.52
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
83.52
Add to cart
Chemical Identifiers
CAS65-85-0
IUPAC Namebenzoic acid
Molecular FormulaC7H6O2
InChI KeyWPYMKLBDIGXBTP-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
Melting point121°C to 124°C
Titration with NaOH>=98.5 %
Water=<0.5 % (K.F.)