3,5-Bis(trifluoromethyl)phenyl isocyanate, 98%, Thermo Scientific Chemicals
3,5-Bis(trifluoromethyl)phenyl isocyanate, 98%, Thermo Scientific Chemicals
3,5-Bis(trifluoromethyl)phenyl isocyanate, 98%, Thermo Scientific Chemicals
3,5-Bis(trifluoromethyl)phenyl isocyanate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3,5-Bis(trifluoromethyl)phenyl isocyanate, 98%, Thermo Scientific Chemicals
CAS: 16588-74-2 | C9H3F6NO | 255.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number ACR314170010
Price (MYR)
502.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
502.00
Add to cart
Chemical Identifiers
CAS536-47-0
IUPAC NameN1,N1-dimethylbenzene-1,4-diamine; sulfuric acid
Molecular FormulaC8H14N2O4S
InChI KeyGLUKPDKNLKRLHX-UHFFFAOYSA-N
SMILESOS(O)(=O)=O.CN(C)C1=CC=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5% (dry wt. basis)
Loss on Drying<5.0% (105?/constant wt.)
Appearance (Color)White to brown or grey