α'-Bromo-α,α,α-trifluoro-p-xylene, 98%, Thermo Scientific Chemicals
α'-Bromo-α,α,α-trifluoro-p-xylene, 98%, Thermo Scientific Chemicals
α'-Bromo-α,α,α-trifluoro-p-xylene, 98%, Thermo Scientific Chemicals
α'-Bromo-α,α,α-trifluoro-p-xylene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
α'-Bromo-α,α,α-trifluoro-p-xylene, 98%, Thermo Scientific Chemicals
CAS: 402-49-3 | C8H6BrF3 | 239.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number ACR312990050
Price (MYR)
599.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
599.00
Add to cart
Chemical Identifiers
CAS17480-69-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colourless to pale yellow
Formliquid
Assay (GC)>98.5%
Optical Rotation-39? ?2? (neat)
Refractive Index1.5625-1.5655