1,2-Diiodotetrafluorobenzene, 99%, Thermo Scientific Chemicals
1,2-Diiodotetrafluorobenzene, 99%, Thermo Scientific Chemicals
1,2-Diiodotetrafluorobenzene, 99%, Thermo Scientific Chemicals
1,2-Diiodotetrafluorobenzene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,2-Diiodotetrafluorobenzene, 99%, Thermo Scientific Chemicals
CAS: 2708-97-6 | C6F4I2 | 401.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number ACR311690010
Price (MYR)
711.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
711.00
Add to cart
Specifications
Chemical Name or Material1,2-Diiodotetrafluorobenzene
CAS2708-97-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee