Octyl acetate, 99+%, Thermo Scientific Chemicals
Octyl acetate, 99+%, Thermo Scientific Chemicals
Octyl acetate, 99+%, Thermo Scientific Chemicals
Octyl acetate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Octyl acetate, 99+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number ACR310630050
Price (MYR)
327.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
327.00
Add to cart
Specifications
Chemical Name or MaterialOctyl acetate
Name Note99+%
CAS112-14-1
CAS Min %99.0
Health Hazard 1